Germanium(IV) ethoxide ≥97%

Supplier: BeanTown Chemical

Synonyms: Germanium tetraethanolate, Germanic acid tetraethyl ester, Tetraethyl orthogermanate, Germanium tetraethoxide, Germanium (IV) ethanolate

128680-1G 128680-25G 128680-5G
BT128680-1GEA 52 USD
BT128680-1G BT128680-25G BT128680-5G
Germanium(IV) ethoxide ≥97%
Germanium(IV) ethoxide

CAS: 14165-55-0; EC No: 238-007-8; MDL No: MFCD00015121 UN No: UN1993; Haz Class: 3; Packing Group: III Liquid; Linear Formula: Ge(OCH2CH3)4; Molecular Formula: C8H20GeO4 ; MW: 252.84 Flash point: 48°C (118°F) Density (g/mL): 1.14 ; Refractive Index: 1.407 Moisture Sensitive


  • Refractive index: 1.407
  • Moisture Sensitive
  • Liquid

Formula: C₈H₂₀GeO₄
MW: 252.85 g/mol
Density: 1.14 g/mL
Flash Pt: 48 °C (118 °F)
MDL Number: MFCD00015121
CAS Number: 14165-55-0
EINECS: 238-007-8
ADR: 3,III

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR