Barium titanate ≥99.9%

Supplier: BeanTown Chemical

Synonyms: Barium titanate, Barium tantalum oxide, Barium titanate (IV), Barium titanium oxide

128640-100G 128640-25G
BT128640-100GEA 394 USD
BT128640-100G BT128640-25G
Barium titanate ≥99.9%
Barium titanate

CAS: 12047-27-7; EC No: 234-975-0; MDL No: MFCD00003447 UN No: UN1564; Haz Class: 6.1; Packing Group: III 3-12mm Pieces; Molecular Formula: BaTiO3; MW: 233.19 Density (g/mL): 6.08


  • 3-12mm Pieces

Formula: BaO₃Ti
MW: 233.21 g/mol
Melting Pt: 1625
Density: 6.08 g/mL
MDL Number: MFCD00003447
CAS Number: 12047-27-7
EINECS: 234-975-0
UN: 1564
ADR: 6.1 III
Merck Index: 12,01029

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR