Barium iodide, hydrate ≥95%
Supplier: BeanTown Chemical
|
Warning
|
128605-100G
128605-25G
BT128605-100GEA
254
USD
BT128605-100G
BT128605-25G
Barium iodide, hydrate ≥95%
Barium iodide
CAS: 13718-50-8; EC No: 237-276-9; MDL No: MFCD00003452
UN No: UN1564; Haz Class: 6.1; Packing Group: III
Powder; Molecular Formula: BaI2·xH2O; MW: 391.15 (anhydrous)
Hygroscopic , Light Sensitive
- Powder
- Hygroscopic, Light Sensitive
Formula:
BaI₂ |
MDL Number:
MFCD00003452 CAS Number: 13718-50-8 EINECS: 237-276-9 UN: 1564 ADR: 6.1,III |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...