Dichloro(phenyl)phosphine ≥97%

Supplier: BeanTown Chemical

Synonyms: P,P-Dichlorophenylphosphine, Phenyldichlorophosphine

128410-1KG 128410-250G 128410-50G
BT128410-1KGEA 167 USD
BT128410-1KG BT128410-250G BT128410-50G
Dichloro(phenyl)phosphine ≥97%
Dichloro(phenyl)phosphine

CAS: 644-97-3; EC No: 211-425-8; MDL No: MFCD00000528; RTECS: TB2478000 UN No: UN2798; Haz Class: 8; Packing Group: II Liquid; Linear Formula: C6H5PCl2; MW: 178.98 Melting Point: -51°; Boiling Point: 222°; Flash point: 110°C (230°F) Density (g/mL): 1.319; Refractive Index: 1.597 Air Sensitive, Moisture Sensitive


  • RTECS: TB2478000
  • Refractive index: 1.597
  • Air Sensitive, Moisture Sensitive
  • Liquid

Formula: C₆H₅Cl₂P
MW: 178.98 g/mol
Boiling Pt: 222 °C
Melting Pt: –51 °C
Density: 1.319 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00000528
CAS Number: 644-97-3
EINECS: 211-425-8
UN: 2798
ADR: 8,II

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR