N,N-Dimethyl-1-dodecylamine N-oxide ≥95%
Supplier: BeanTown Chemical
CAS: 1643-20-5; EC No: 216-700-6; MDL No: MFCD00002049; RTECS: JR6650000
UN No: UN3259; Haz Class: 8; Packing Group: III
Solid; Linear Formula: CH3(CH2)11N(O)(CH3)2; Molecular Formula: C14H31NO ; MW: 229.40
Melting Point: 132-133°
Density (g/mL): 0.996
- RTECS: JR6650000
- Solid
Formula:
C₁₄H₃₁NO Melting Pt: 132…133 °C Density: 0.996 g/mL |
MDL Number:
MFCD00002049 CAS Number: 1643-20-5 EINECS: 216-700-6 ADR: 8,III |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...