Potassium tetrachloroaurate hydrate ≥99.99% (trace metals basis)

Supplier: BeanTown Chemical

Synonyms: Gold potassium chloride hydrate, Potassium tetrachloroaurate(III) hydrate, Potassium gold(III) chloride hydrate

128365-1G 128365-5G
BT128365-1GEA 125.28 USD
BT128365-1G BT128365-5G
Potassium tetrachloroaurate hydrate ≥99.99% (trace metals basis)
Potassium tetrachloroaurate hydrate

CAS: 27988-75-6; EC No: 237-190-1; MDL No: MFCD00043088 UN No: UN3260; Haz Class: 8; Packing Group: III Crystalline; Linear Formula: KAuCl4·xH2O; MW: 377.88 (anhydrous) Melting Point: 300°


  • Crystalline

Formula: AuCl₄K·xH₂O
MW: 377.88 g/mol (anhy)
Melting Pt: 300 °C
MDL Number: MFCD00043088
CAS Number: 27988-75-6
EINECS: 237-190-1
UN: 3260
ADR: 8 III

Order Now

Specification Test Results

Assay 99.99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR