tert-Pentylamine ≥98%
Supplier: BeanTown Chemical
CAS: 594-39-8; EC No: 209-839-9; MDL No: MFCD00008056
UN No: UN1106; Haz Class: 3 (8); Packing Group: II
Liquid; Linear Formula: CH3CH2C(CH3)2NH2; Molecular Formula: C5H13N; MW: 87.17
Boiling Point: 77°; Flash point: -1°C (30°F)
Density (g/mL): 0.746 ; Refractive Index: 1.400
- Refractive index: 1.400
- Liquid
Formula:
C₅H₁₃N Boiling Pt: 77 °C Density: 0.746 g/mL Flash Pt: –1 °C (30 °F) |
MDL Number:
MFCD00008056 CAS Number: 594-39-8 EINECS: 209-839-9 UN: 1106 ADR: 3,II |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...