4-Morpholinoaniline ≥98%
Supplier: BeanTown Chemical
Synonyms:
4-(4-Morpholinyl)aniline
128305-1G
128305-25G
128305-5G
BT128305-1GEA
41
USD
BT128305-1G
BT128305-25G
BT128305-5G
4-Morpholinoaniline ≥98%
4-Morpholinoaniline
CAS: 2524-67-6; EC No: 219-760-1; MDL No: MFCD00006169
UN No: UN2811; Haz Class: 6.1; Packing Group: III
Crystalline/Powder; Molecular Formula: C10H14N2O; MW: 178.24
Melting Point: 132-135°
- Crystalline/Powder
Formula:
C₁₀H₁₄N₂O MW: 178.23 g/mol Melting Pt: 132…135 °C |
MDL Number:
MFCD00006169 CAS Number: 2524-67-6 EINECS: 219-760-1 UN: 2811 ADR: 6.1,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...