Phenylphosphine
Supplier: BeanTown Chemical
CAS: 638-21-1; EC No: 211-325-4; MDL No: MFCD00002094
UN No: UN2845; Haz Class: 4.2; Packing Group: I
Liquid; Linear Formula: C6H5PH2; MW: 110.10
Boiling Point: 160°; Flash point: -26°C (-15°F)
Density (g/mL): 1.001; Refractive Index: 1.578
Air Sensitive, Moisture Sensitive
- Refractive index: 1.578
- Air Sensitive, Moisture Sensitive
- Liquid
Formula:
C₆H₇P Boiling Pt: 160 °C Density: 1.001 g/mL Flash Pt: –26 °C (–15 °F) |
MDL Number:
MFCD00002094 CAS Number: 638-21-1 EINECS: 211-325-4 UN: 2845 ADR: 4.2,I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...