Lithium nitrate hydrate ≥99.999% (trace metals basis)
Supplier: BeanTown Chemical
|
Danger
|
128230-10G
128230-50G
BT128230-10GEA
61.78
USD
BT128230-10G
BT128230-50G
Lithium nitrate hydrate ≥99.999% (trace metals basis)
Lithium nitrate hydrate
CAS: 7790-69-4; EC No: 232-218-9; MDL No: MFCD00011094
UN No: UN2722; Haz Class: 5.1; Packing Group: III
Crystalline Aggregates; Molecular Formula: LiNO3·xH2O; MW: 68.95 (anhydrous)
Hygroscopic
- Crystalline Aggregates
- Hygroscopic
Formula:
LiNO₃·H₂O MW: 68.95 g/mol (anhy) Storage Temperature: Ambient |
MDL Number:
MFCD00011094 CAS Number: 7790-69-4 EINECS: 232-218-9 UN: 2722 ADR: 5.1,III Merck Index: 14,5536 |
Specification Test Results
Assay | 99.999% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...