Benzonitrile ≥99%

Supplier: BeanTown Chemical

128205-100G 128205-2.5KG 128205-500G
BT128205-100GEA 39.04 USD
BT128205-100G BT128205-2.5KG BT128205-500G
Benzonitrile ≥99%
Benzonitrile

CAS: 100-47-0; EC No: 202-855-7; MDL No: MFCD00001770; RTECS: DI2450000 UN No: UN2224; Haz Class: 6.1; Packing Group: II Liquid; Molecular Formula: C7H5N; MW: 103.12 Melting Point: -13°; Boiling Point: 191°; Flash point: 70°C (158°F) Refractive Index: 1.528


  • RTECS: DI2450000
  • Refractive index: 1.528
  • Liquid

Formula: C₇H₅N
Boiling Pt: 191 °C
Melting Pt: –13 °C
Flash Pt: 70 °C (158 °F)
MDL Number: MFCD00001770
CAS Number: 100-47-0
EINECS: 202-855-7
UN: 2224
ADR: 6.1,II

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR