Cadmium sulfide ≥99.99% (trace metals basis)
Supplier: BeanTown Chemical
Synonyms:
Cadmium (II) sulphide
128180-100G
128180-25G
BT128180-100GEA
776
USD
BT128180-100G
BT128180-25G
Cadmium sulfide ≥99.99% (trace metals basis)
Cadmium sulphide
CAS: 1306-23-6; EC No: 215-147-8; MDL No: MFCD00010922; RTECS: EV3150000
UN No: UN3077; Haz Class: 9; Packing Group: III
3-12mm Sintered Lump ; Molecular Formula: CdS; MW: 144.48
Melting Point: 980° (sublimes)
Density (g/mL): 4.82
- RTECS: EV3150000
- 3-12mm Sintered Lump
Formula:
CdS MW: 144.48 g/mol Melting Pt: 980 °C (subl.) Density: 4.82 g/mL |
MDL Number:
MFCD00010922 CAS Number: 1306-23-6 EINECS: 215-147-8 UN: 3077 ADR: 9,III |
Specification Test Results
Assay | 99.99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...