2,6-Dichloroaniline ≥98%

Supplier: BeanTown Chemical
Danger

128125-100G 128125-25G
BT128125-100GEA 83 USD
BT128125-100G BT128125-25G
2,6-Dichloroaniline ≥98%
2,6-Dichloroaniline

CAS: 608-31-1; EC No: 210-160-5; MDL No: MFCD00007675; RTECS: CX9862655 UN No: UN3442; Haz Class: 6.1; Packing Group: II Crystalline/Solid; Linear Formula: (Cl)2C6H3NH2; Molecular Formula: C6H5Cl2N; MW: 162.02 Melting Point: 37-40°; Boiling Point: 97°/5 mmHg; Flash point: 118°C (244°F) Density (g/mL): 1.280


  • RTECS: CX9862655
  • Crystalline/Solid

Formula: C₆H₅Cl₂N
MW: 162.02 g/mol
Boiling Pt: 97 °C (5 mmHg)
Melting Pt: 37…40 °C
Density: 1.280 (g/mL)
Flash Pt: 118 °C
MDL Number: MFCD00007675
CAS Number: 608-31-1
EINECS: 210-160-5
UN: 3442
ADR: 6.1,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR