Iron(II) bromide, anhydrous ≥98%

Supplier: BeanTown Chemical
Danger

Synonyms: Iron dibromide, Ferrous bromide, Iron(II) bromide

128120-25G 128120-5G
BT128120-25GEA 205 USD
BT128120-25G BT128120-5G
Iron(II) bromide, anhydrous ≥98%
Iron(II) bromide

CAS: 7789-46-0; EC No: 232-168-8; MDL No: MFCD00016081 UN No: UN3260; Haz Class: 8; Packing Group: III Granular; Linear Formula: Br2Fe; MW: 215.67 Air Sensitive , Hygroscopic


  • Granular
  • Air Sensitive , Hygroscopic

Formula: Br₂Fe
MW: 215.66 g/mol
MDL Number: MFCD00016081
CAS Number: 7789-46-0
EINECS: 232-168-8
UN: 3260
ADR: 8 III

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR