Holmium ≥99.9% (trace rare earth metals basis), powder -325 mesh
Supplier: BeanTown Chemical
|
Danger
|
128025-100G
128025-25G
BT128025-100GEA
781
USD
BT128025-100G
BT128025-25G
Holmium ≥99.9% (trace rare earth metals basis), powder -325 mesh
Holmium
CAS: 7440-60-0; EC No: 231-169-0; MDL No: MFCD00011049
UN No: UN1383; Haz Class: 4.2; Packing Group: I
Powder; Molecular Formula: Ho; MW: 164.93
Melting Point: 1474°; Boiling Point: 2695°
Density (g/mL): 8.8
Formula:
Ho MW: 164.93 g/mol Boiling Pt: 2695 °C Melting Pt: 1474 °C Density: 8.8 g/cm³ |
MDL Number:
MFCD00011049 CAS Number: 7440-60-0 EINECS: 231-169-0 UN: 1383 ADR: 4.2,II |
Specification Test Results
Assay | 99.9% |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...