Bromoacetic acid ≥98%
Supplier: BeanTown Chemical
127960-1KG
127960-250G
127960-5KG
BT127960-1KGEA
98.64
USD
BT127960-1KG
BT127960-250G
BT127960-5KG
Bromoacetic acid ≥98%
Bromoacetic acid
CAS: 79-08-3; EC No: 201-175-8; MDL No: MFCD00002678; RTECS: AF5950000
UN No: UN3425; Haz Class: 8; Packing Group: II
Crystalline; Linear Formula: BrCH2COOH; MW: 138.95
Melting Point: 46-49°; Boiling Point: 208°; Flash point:
- RTECS: AF5950000
- Hygroscopic, Light Sensitive
- Crystalline
Formula:
BrCH₂COOH Boiling Pt: 208 °C Melting Pt: 46…49 °C Density: 1.93 g/mL Flash Pt: >110 °C (>230 °F) |
MDL Number:
MFCD00002678 CAS Number: 79-08-3 EINECS: 201-175-8 UN: 3425 ADR: 8,II |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...