2,2'-Bipyridyl ≥98.0% ACS

Supplier: BeanTown Chemical

Synonyms: α,α′-Dipyridyl, 2,2'-Bipyridine, 2,2'-Dipyridyl

127950-10G 127950-250G 127950-50G
BT127950-10GEA 28.26 USD
BT127950-10G BT127950-250G BT127950-50G
2,2'-Bipyridyl ≥98.0% ACS
2,2'-Bipyridyl

CAS: 366-18-7; EC No: 206-674-4; MDL No: MFCD00006212; RTECS: DW1750000 UN No: UN2811; Haz Class: 6.1; Packing Group: III Crystalline; Molecular Formula: C10H8N2; MW: 156.18 Melting Point: 70-73°; Boiling Point: 273°


  • RTECS: DW1750000
  • Crystalline

Formula: C₁₀H₈N₂
MW: 156.19 g/mol
Boiling Pt: 273 °C
Melting Pt: 70…73 °C
MDL Number: MFCD00006212
CAS Number: 366-18-7
EINECS: 206-674-4
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e86bbc29edb6c5d09c058b8efcb6b9eb7f4d070c&vid=9e3a03e37045643109ae0765802e1997d30604ae&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 98.0% min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR