3,5-Dichlorophenyl isocyanate ≥97%

Supplier: BeanTown Chemical

Synonyms: 3,5-Dichlorophenylisocyantae

127915-25G 127915-5G
BT127915-25GEA 277 USD
BT127915-25G BT127915-5G
3,5-Dichlorophenyl isocyanate ≥97%
3,5-Dichlorophenyl isocyanate

CAS: 34893-92-0; EC No: 252-276-9; MDL No: MFCD00013859 UN No: UN2250; Haz Class: 6.1; Packing Group: II Solid; Molecular Formula: C7H3Cl2NO; MW: 188.01 Melting Point: 32-34°; Flash point: 110°C (230°F) Density (g/mL): 1.38 Moisture Sensitive


  • Moisture Sensitive
  • Solid

Formula: C₇H₃Cl₂NO
MW: 188.01 g/mol
Melting Pt: 32…34 °C
Density: 1.38 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00013859
CAS Number: 34893-92-0
EINECS: 252-276-9
UN: 2250
ADR: 6.1,II

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR