4-Bromo-o-toluidine ≥98%

Supplier: BeanTown Chemical

Synonyms: 2-Amino-5-bromotoluene, 4-Bromo-2-methylaniline

127880-100G 127880-25G
BT127880-100GEA 172 USD
BT127880-100G BT127880-25G
4-Bromo-o-toluidine ≥98%
4-Bromo-o-toluidine

CAS: 583-75-5; EC No: 209-519-9; MDL No: MFCD00007825; RTECS: XU4250000 UN No: UN2811; Haz Class: 6.1; Packing Group: III Powder; Molecular Formula: C7H8BrN; MW: 186.05 Melting Point: 57-59°; Boiling Point: 240°; Flash point: 110°C (230°F)


  • RTECS: XU4250000
  • Powder

Formula: C₇H₈BrN
MW: 186.05 g/mol
Boiling Pt: 240 °C
Melting Pt: 57…59 °C
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00007825
CAS Number: 583-75-5
EINECS: 209-519-9
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR