Hexaammineruthenium trichloride

Supplier: BeanTown Chemical

Synonyms: Hexaammineruthenium(III) chloride, Hexaammineruthenium (III) chloride

127875-1G 127875-250MG 127875-5G
BT127875-1GEA 144.04 USD
BT127875-1G BT127875-250MG BT127875-5G
Hexaammineruthenium trichloride
Hexaammineruthenium trichloride

CAS: 14282-91-8; EC No: 238-176-8; MDL No: MFCD00011478; RTECS: VM2651530 UN No: UN3288; Haz Class: 6.1; Packing Group: III Powder; Linear Formula: Ru(NH3)6Cl3; Molecular Formula: H18Cl3N6Ru; MW: 309.61


  • RTECS: VM2651530
  • Powder

Formula: H₁₈Cl₃N₆Ru
MW: 309.61 g/mol
MDL Number: MFCD00011478
CAS Number: 14282-91-8
EINECS: 238-176-8
UN: 3288
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR