Ruthenium ≥99.9% (trace metals basis), powder -325 mesh

Supplier: BeanTown Chemical

127835-1G 127835-5G
BT127835-1GEA 121.16 USD
BT127835-1G BT127835-5G
Ruthenium ≥99.9% (trace metals basis), powder -325 mesh
Ruthenium

CAS: 7440-18-8; EC No: 231-127-1; MDL No: MFCD00011207 UN No: UN3089; Haz Class: 4.1; Packing Group: III Powder; Molecular Formula: Ru; MW: 101.07 Melting Point: 2310°; Boiling Point: 3900° Density (g/mL): 12.45


  • Powder

Formula: Ru
MW: 101.07
Boiling Pt: 3900 °C
Melting Pt: 2310 °C
Density: 12.45 g/mL
MDL Number: MFCD00011207
CAS Number: 7440-18-8
EINECS: 231-127-1
UN: 3089
ADR: 4.1 III

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR