2,2':6',2''-Terpyridine ≥97%

Supplier: BeanTown Chemical

Synonyms: TERPY

127765-1G 127765-250MG 127765-5G
BT127765-1GEA 161.42 USD
BT127765-1G BT127765-250MG BT127765-5G
2,2':6',2''-Terpyridine ≥97%
2,2':6',2''-Terpyridine

CAS: 1148-79-4; EC No: 214-559-5; MDL No: MFCD00006213 UN No: UN2811; Haz Class: 6.1; Packing Group: I Crystalline; Molecular Formula: C15H11N3; MW: 233.27 Melting Point: 89-91°


  • Crystalline

Formula: C₁₅H₁₁N₃
MW: 233.27 g/mol
Melting Pt: 89…91 °C
MDL Number: MFCD00006213
CAS Number: 1148-79-4
EINECS: 214-559-5
ADR: 6.1,I

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR