Ytterbium ≥99.9% (trace rare earth metals basis), powder -40 mesh

Supplier: BeanTown Chemical

127740-1G 127740-5G
BT127740-1GEA 53 USD
BT127740-1G BT127740-5G
Ytterbium ≥99.9% (trace rare earth metals basis), powder -40 mesh
Ytterbium

CAS: 7440-64-4; EC No: 231-173-2; MDL No: MFCD00011286; RTECS: ZG1925000 UN No: UN3089; Haz Class: 4.1; Packing Group: II Powder, packaged under argon in glass bottle; Molecular Formula: Yb; MW: 173.04 Melting Point: 824°; Boiling Point: 1196° Density (g/mL): 6.54


  • RTECS: ZG1925000
  • Powder, packaged under argon in glass bottle

Formula: Yb
MW: 173.04 g/mol
Boiling Pt: 1196 °C
Melting Pt: 824 °C
Density: 6.54 g/mL
MDL Number: MFCD00011286
CAS Number: 7440-64-4
EINECS: 231-173-2
UN: 3089
ADR: 4.1,III

Order Now

Specification Test Results

Assay 99.9%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR