Bis(2-bromoethyl)ether ≥96%
Supplier: BeanTown Chemical
CAS: 5414-19-7; EC No: 226-504-2; MDL No: MFCD00039196
UN No: UN2810; Haz Class: 6.1; Packing Group: II
Liquid; Linear Formula: BrCH2CH2OCH2CH2Br; Molecular Formula: C4H8Br2O; MW: 231.91
Boiling Point: 92-93°/12 mmHg; Flash point: 85°C (185°F)
Density (g/mL): 1.845; Refractive Index: 1.514
- Refractive index: 1.514
- Liquid
Formula:
C₄H₈Br₂O Boiling Pt: 92…93 °C/12 mmHg Density: 1.845 g/mL Flash Pt: 85 °C (185 °F) |
MDL Number:
MFCD00039196 CAS Number: 5414-19-7 EINECS: 226-504-2 UN: 2810 ADR: 6.1,II |
Specification Test Results
Assay | 96% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...