2-Bromoaniline ≥98%

Supplier: BeanTown Chemical

127695-100G 127695-25G 127695-500G
BT127695-100GEA 184 USD
BT127695-100G BT127695-25G BT127695-500G
2-Bromoaniline ≥98%
2-Bromoaniline

CAS: 615-36-1; EC No: 210-421-3; MDL No: MFCD00007632 UN No: UN2811; Haz Class: 6.1; Packing Group: III Solid; Molecular Formula: C6H6BrN; MW: 172.02 Melting Point: 24-28°; Boiling Point: 229° Density (g/mL): 1.52


  • Solid

Formula: C₆H₆BrN
MW: 172.02 g/mol
Boiling Pt: 229 °C
Melting Pt: 24…28 °C
Density: 1.52 g/mL
MDL Number: MFCD00007632
CAS Number: 615-36-1
EINECS: 210-421-3
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR