Triphenylphosphine dibromide ≥96%

Supplier: BeanTown Chemical

Synonyms: Dibromotriphenylphosphorane

127690-10G 127690-250G 127690-50G
BT127690-10GEA 41 USD
BT127690-10G BT127690-250G BT127690-50G
Triphenylphosphine dibromide ≥96%
Triphenylphosphine dibromide

CAS: 1034-39-5; EC No: 213-855-1; MDL No: MFCD00000054 UN No: UN3261; Haz Class: 8; Packing Group: II Crystalline; Linear Formula: (C6H5)3PBr2; Molecular Formula: C18H15Br2P; MW: 422.10 Melting Point: 235° (decomposes) Moisture Sensitive


  • Moisture Sensitive
  • Crystalline

Formula: C₁₈H₁₅Br₂P
MW: 422.1
Melting Pt: 235 °C (dec.)
MDL Number: MFCD00000054
CAS Number: 1034-39-5
EINECS: 213-855-1
UN: 3261
ADR: 8 II

Order Now

Specification Test Results

Assay 96%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR