Phosphorus, red ≥99.999%, lumps
Supplier: BeanTown Chemical
127660-10G
127660-1G
127660-50G
BT127660-10GEA
199.49
USD
BT127660-10G
BT127660-1G
BT127660-50G
Phosphorus, red ≥99.999%, lumps
Phosphorus
CAS: 7723-14-0; EC No: 231-768-7; MDL No: MFCD00133771; RTECS: TH3495000
UN No: UN1338; Haz Class: 4.1 ; Packing Group: III
Solid; Molecular Formula: P; MW: 30.97
Melting Point: 416° (sublimes)
Density (g/mL): 2.34
- RTECS: TH3495000
- Solid
Formula:
P MW: 30.97 g/mol Melting Pt: 416 °C (subl.) Density: 2.34 g/mL |
MDL Number:
MFCD00133771 CAS Number: 7723-14-0 EINECS: 231-768-7 UN: 1338 ADR: 4.1 III |
Specification Test Results
Assay | 99.999% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...