Hydrazinium hydroxide ≥99%
Supplier: BeanTown Chemical
Synonyms:
Hydrazine hydrate, Hydrazine monohydrate
127575-100G
127575-2KG
127575-500G
BT127575-100GEA
48.51
USD
BT127575-100G
BT127575-2KG
BT127575-500G
Hydrazinium hydroxide ≥99%
Hydrazinium hydroxide
CAS: 7803-57-8; EC No: 206-114-9; MDL No: MFCD00149931; RTECS: MV4590000
UN No: UN2030; Haz Class: 8 (6.1); Packing Group: II
Liquid; Linear Formula: H2NNH2·H2O; MW: 50.06
Melting Point: -52°; Boiling Point: 120.1°; Flash point: 75°C (167°F)
Density (g/mL): 1.032; Refractive Index: 1.428
- RTECS: MV4590000
- Refractive index: 1.428
- Liquid
Formula:
NH₂NH₂·H₂O MW: 50.06 g/mol Boiling Pt: 120.1 °C Melting Pt: –52 °C Density: 1.032 g/mL Flash Pt: 75 °C (167 °F) |
MDL Number:
MFCD00149931 CAS Number: 7803-57-8 EINECS: 206-114-9 UN: 2030 ADR: 8,II |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...