Ethyl-(S)-(-)-lactate ≥99%
Supplier: BeanTown Chemical
CAS: 687-47-8; EC No: 211-694-1; MDL No: MFCD00065359; RTECS: OD5070000
UN No: UN1192; Haz Class: 3; Packing Group: III
Liquid; Molecular Formula: C5H10O3; MW: 118.13
Boiling Point: 152-156°; Flash point: 46°C (114°F)
Density (g/mL): 1.036; Refractive Index: 1.413; Optical Rotation: [α]20/D -10.5±1°, neat
- RTECS: OD5070000
- Optical rotation: [α]20/D -10.5±1°, neat
- Refractive index: 1.413
- Liquid
Formula:
C₅H₁₀O₃ Boiling Pt: 152…156 °C Density: 1.036 g/mL Flash Pt: 46 °C (114 °F) |
MDL Number:
MFCD00004518 CAS Number: 687-47-8 EINECS: 211-694-1 UN: 1192 ADR: 3,III |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...