2-Amino-6-methylpyridine ≥99%

Supplier: BeanTown Chemical

Synonyms: 6-Methyl-2-pyridylamine, 6-Methylpyridin-2-amine, 2-Amino-6-picoline

127460-100G 127460-500G
BT127460-100GEA 26 USD
BT127460-100G BT127460-500G
2-Amino-6-methylpyridine ≥99%
2-Amino-6-methylpyridine

CAS: 1824-81-3; EC No: 217-360-1; MDL No: MFCD00006331; RTECS: US1885000 UN No: UN2811; Haz Class: 6.1; Packing Group: II Crystalline; Molecular Formula: C6H8N2 ; MW: 108.14 Melting Point: 40-44°; Boiling Point: 208-209°; Flash point: 103°C (217°F) Hygroscopic


  • RTECS: US1885000
  • Hygroscopic
  • Crystalline

Formula: C₆H₈N₂
MW: 108.14 g/mol
Boiling Pt: 214.4
Melting Pt: 40…44 °C
Flash Pt: 103 °C (217 °F)
MDL Number: MFCD00006331
CAS Number: 1824-81-3
EINECS: 217-360-1
UN: 2811
ADR: 6.1 II

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR