Tin selenide ≥99.999% (trace metals basis)
Supplier: BeanTown Chemical
127435-25G
127435-5G
BT127435-25GEA
367
USD
BT127435-25G
BT127435-5G
Tin selenide ≥99.999% (trace metals basis)
Tin selenide
CAS: 1315-06-6; EC No: 215-257-6; MDL No: MFCD00054049
UN No: UN3077; Haz Class: 9; Packing Group: III
Pieces; Linear Formula: SnSe; MW: 197.65
Melting Point: 861°
Density (g/mL): 6.18
- Pieces
Formula:
SeSn MW: 197.67 g/mol Melting Pt: 861 °C Density: 6.18 g/mL |
MDL Number:
MFCD00054049 CAS Number: 1315-06-6 EINECS: 215-257-6 ADR: 9,III |
Specification Test Results
Assay | 99.999% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...