Pyridine-2-carbaldehyde ≥99%

Supplier: BeanTown Chemical

Synonyms: 2-Pyridinecarboxaldehyde, 2-Pyridinecarbaldehyde, Pyridine-2-carboxaldehyde

127325-100G 127325-25G 127325-500G
BT127325-100GEA 65 USD
BT127325-100G BT127325-25G BT127325-500G
Pyridine-2-carbaldehyde ≥99%
Pyridine-2-carbaldehyde

CAS: 1121-60-4; EC No: 214-333-6; MDL No: MFCD00006290 UN No: UN2810; Haz Class: 6.1; Packing Group: III Liquid; Molecular Formula: C6H5NO ; MW: 107.11 Melting Point: -21°; Boiling Point: 180-181°; Flash point: 68°C (154°F) Density (g/mL): 1.126; Refractive Index: 1.536


  • Refractive index: 1.536
  • Liquid

Formula: C₆H₅NO
Boiling Pt: 180…181 °C
Melting Pt: –21 °C
Density: 1.126 g/mL
Flash Pt: 68 °C (154 °F)
MDL Number: MFCD00006290
CAS Number: 1121-60-4
EINECS: 214-333-6
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR