(±)-Methyl-2-bromobutyrate ≥97%
Supplier: BeanTown Chemical
CAS: 3196-15-4; EC No: 221-699-0; MDL No: MFCD00009666
UN No: UN1760; Haz Class: 8; Packing Group: III
Liquid; Linear Formula: CH3CH2CHBrCOOCH3; Molecular Formula: C5H9BrO2 ; MW: 181.03
Boiling Point: 137-138°/50 mmHg; Flash point: 68°C (154°F)
Density (g/mL): 1.573; Refractive Index: 1.452
- Refractive index: 1.452
- Liquid
Formula:
C₅H₉BrO₂ Boiling Pt: 137…138 °C/50 mmHg Density: 1.573 g/mL Flash Pt: 68 °C (154 °F) |
MDL Number:
MFCD00009666 CAS Number: 3196-15-4 EINECS: 221-699-0 ADR: 8,III |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...