Samarium(III) fluoride, anhydrous ≥99.99% (trace rare earth metals basis)

Supplier: BeanTown Chemical

127295-1G 127295-25G 127295-5G
BT127295-1GEA 23 USD
BT127295-1G BT127295-25G BT127295-5G
Samarium(III) fluoride, anhydrous ≥99.99% (trace rare earth metals basis)
Samarium(III) fluoride

CAS: 13765-24-7; EC No: 237-367-3; MDL No: MFCD00049540 UN No: UN3288; Haz Class: 6.1; Packing Group: III Powder; Linear Formula: SmF3; MW: 207.35 Melting Point: 1306°; Boiling Point: 2323° Density (g/mL): 6.28 Hygroscopic


  • Hygroscopic
  • Powder

Formula: F₃Sm
MW: 207.36 g/mol
Boiling Pt: 2323 °C
Melting Pt: 1306 °C
Density: 6.28 g/mL
MDL Number: MFCD00049540
CAS Number: 13765-24-7
EINECS: 237-367-3
UN: 3288
ADR: 6.1,III

Order Now

Specification Test Results

Assay 99.99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR