Calcium nitride ≥99%

Supplier: BeanTown Chemical

127245-10G 127245-250G 127245-50G
BT127245-10GEA 82.6 USD
BT127245-10G BT127245-250G BT127245-50G
Calcium nitride ≥99%
Calcium nitride

CAS: 12013-82-0; EC No: 234-592-9; MDL No: MFCD00015985 UN No: UN3178; Haz Class: 4.1; Packing Group: II 12mm & Down Pieces; Molecular Formula: Ca3N2; MW: 148.25 Density (g/mL): 2.63 Moisture Sensitive


  • Moisture Sensitive
  • 12mm & Down Pieces

Formula: Ca₃N₂
Density: 2.63 g/mL
MDL Number: MFCD00015985
CAS Number: 12013-82-0
EINECS: 234-592-9
UN: 3178
ADR: 4.1,II

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR