1-Octyne ≥98%

Supplier: BeanTown Chemical

127240-100G 127240-10G 127240-25G
BT127240-100GEA 170 USD
BT127240-100G BT127240-10G BT127240-25G
1-Octyne ≥98%
1-Octyne

CAS: 629-05-0; EC No: 211-069-3; MDL No: MFCD00009546; RTECS: RI2655000 UN No: UN3295; Haz Class: 3; Packing Group: II Liquid; Linear Formula: CH3(CH2)5C≡CH; Molecular Formula: C8H14; MW: 110.20 Melting Point: -80°; Boiling Point: 127-128°; Flash point: 16°C (61°F) Density (g/mL): 0.747; Refractive Index: 1.416


  • RTECS: RI2655000
  • Refractive index: 1.416
  • Liquid

Formula: C₈H₁₄
MW: 110.2 g/mol
Boiling Pt: 127…128 °C
Melting Pt: –80 °C
Density: 0.747 g/mL
Flash Pt: 16 °C (61 °F)
MDL Number: MFCD00009546
CAS Number: 629-05-0
EINECS: 211-069-3
UN: 3295
ADR: 3,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR