4-Nitro-2,3-lutidine-N-oxide ≥97%

Supplier: BeanTown Chemical

Synonyms: 2,3-Dimethyl-4-nitropyridine 1-oxide, 2,3-Dimethyl-4-nitropyridine N-oxide

127180-1G 127180-25G 127180-5G
BT127180-1GEA 29 USD
BT127180-1G BT127180-25G BT127180-5G
4-Nitro-2,3-lutidine-N-oxide ≥97%
4-Nitro-2,3-lutidine N-oxide

CAS: 37699-43-7; MDL No: MFCD00065172; RTECS: UT2807000 UN No: UN2811; Haz Class: 6.1; Packing Group: III Solid; Molecular Formula: C7H8N2O3; MW: 168.15 Melting Point: 94-98°


  • RTECS: UT2807000
  • Solid

Formula: C₇H₈N₂O₂
MW: 168.15 g/mol
Melting Pt: 94…98 °C
MDL Number: MFCD00065172
CAS Number: 37699-43-7
ADR: 6.1,III

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR