Trichlorosilane ≥98%

Supplier: BeanTown Chemical

127175-100G 127175-25G 127175-500G
BT127175-100GEA 62.33 USD
BT127175-100G BT127175-25G BT127175-500G
Trichlorosilane ≥98%
Trichlorosilane

CAS: 10025-78-2; EC No: 233-042-5; MDL No: MFCD00011518; RTECS: VV5950000 UN No: UN1295; Haz Class: 4.3 (3,8); Packing Group: I Liquid; Linear Formula: SiHCl3; MW: 135.45 Melting Point: -127°; Boiling Point: 32-34°; Flash point: -51°C (-60°F) Density (g/mL): 1.342; Refractive Index: 1.402 Air Sensitive, Moisture Sensitive


  • RTECS: VV5950000
  • Refractive index: 1.402
  • Air Sensitive, Moisture Sensitive
  • Liquid

Formula: HCl₃Si
MW: 135.45 g/mol
Boiling Pt: 32…34 °C
Melting Pt: –127 °C
Density: 1.342 g/mL
Flash Pt: –51 °C (–60 °F)
MDL Number: MFCD00011518
CAS Number: 10025-78-2
EINECS: 233-042-5
UN: 1295
ADR: 4.3,I

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR