Cyclobutanecarbonitrile ≥97%
Supplier: BeanTown Chemical
127170-100G
127170-25G
127170-5G
BT127170-100GEA
1912
USD
BT127170-100G
BT127170-25G
BT127170-5G
Cyclobutanecarbonitrile ≥97%
Cyclobutanecarbonitrile
CAS: 4426-11-3; EC No: 224-606-1; MDL No: MFCD00013735
UN No: UN1992; Haz Class: 3 (6.1); Packing Group: III
Liquid; Molecular Formula: C5H7N; MW: 81.12
Boiling Point: 144-147°; Flash point: 42°C (107°F)
Density (g/mL): 0.868
- Liquid
Formula:
C₅H₇N MW: 81.12 g/mol Boiling Pt: 144…147 °C Density: 0.868 g/mL Flash Pt: 42 °C (107 °F) |
MDL Number:
MFCD00013735 CAS Number: 4426-11-3 EINECS: 224-606-1 UN: 1992 ADR: 3,III |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...