Cesium hydroxide 50% (w/w) in water , 99% (trace metals basis)
Supplier: BeanTown Chemical
127130-C100G
127130-C500G
BT127130-C100GEA
116.09
USD
BT127130-C100G
BT127130-C500G
Cesium hydroxide 50% (w/w) in water , 99% (trace metals basis)
Cesium hydroxide
CAS: 21351-79-1; EC No: 244-344-1; MDL No: MFCD00010964
UN No: UN2681; Haz Class: 8; Packing Group: II
Liquid; Linear Formula: CsOH; MW: 149.91
Density (g/mL): 1.72
Air Sensitive
- Air Sensitive
- Liquid
Formula:
CsOH MW: 149.91 g/mol Density: 1.72 g/mL |
MDL Number:
MFCD00010964 CAS Number: 21351-79-1 EINECS: 244-344-1 UN: 2681 ADR: 8,II |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...