3-Hepten-2-one ≥97%

Supplier: BeanTown Chemical

Synonyms: Hept-3-en-2-one

127120-10G 127120-50G
BT127120-10GEA 54 USD
BT127120-10G BT127120-50G
3-Hepten-2-one ≥97%
3-Hepten-2-one

CAS: 1119-44-4; EC No: 214-278-8; MDL No: MFCD00015564 UN No: UN1224; Haz Class: 3; Packing Group: III Liquid; Molecular Formula: C7H12O; MW: 112.16 Flash point: 49°C (120°F) Density (g/mL): 0.86


  • Liquid

Formula: C₇H₁₂O
MW: 112.17 g/mol
Density: 0.86 g/mL
Flash Pt: 49 °C (120 °F)
MDL Number: MFCD00015564
CAS Number: 1119-44-4
EINECS: 214-278-8
ADR: 3 III

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR