Dibutyl sulphide ≥98%

Supplier: BeanTown Chemical

Synonyms: Dibutylsulphide, Butyl sulphide, n-Butyl sulphide

127095-100ML 127095-500ML
BT127095-100MLEA 43 USD
BT127095-100ML BT127095-500ML
Dibutyl sulphide ≥98%
Dibutyl sulphide

CAS: 544-40-1; EC No: 208-870-5; MDL No: MFCD00009468; RTECS: ER6417000 UN No: UN3334; Haz Class: 9; Packing Group: III Liquid; Linear Formula: CH3(CH2)3S(CH2)3CH3; Molecular Formula: C8H18S; MW: 146.29 Melting Point: -76°; Boiling Point: 188-189°; Flash point: 77°C (171°F) Density (g/mL): 0.838; Refractive Index: 1.453


  • RTECS: ER6417000
  • Refractive index: 1.453
  • Liquid

Formula: C₈H₁₈S
MW: 146.3 g/mol
Boiling Pt: 189
Melting Pt: -80
Density: 0.838 g/mL
Flash Pt: 77 °C (171 °F)
MDL Number: MFCD00009468
CAS Number: 544-40-1
EINECS: 208-870-5
UN: 3334
ADR: 9 III
Merck Index: 12,01626

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR