Dichloromethane-[D₂] ≥100% (Isotopic)

Supplier: BeanTown Chemical

127060-10EACH 127060-2EACH
BT127060-10EAEA 432.33 USD
BT127060-10EA BT127060-2EACH
Dichloromethane-[D₂] ≥100% (Isotopic)
Dichloromethane-[D₂]

CAS: 1665-00-5; EC No: 216-776-0; MDL No: MFCD00000882 UN No: UN1593; Haz Class: 6.1; Packing Group: III Liquid, in prescored ampules, 0.75ml/ampule; Molecular Formula: CD2Cl2; MW: 86.95 Boiling Point: 40° Density (g/mL): 1.362; Refractive Index: 1.422 Moisture Sensitive


  • Refractive index: 1.422
  • Moisture Sensitive
  • Liquid, in prescored ampules, 0.75ml/ampule

Formula: CD₂Cl₂
Boiling Pt: 40 °C
Density: 1.362 g/mL
MDL Number: MFCD00000882
CAS Number: 1665-00-5
EINECS: 216-776-0
UN: 1593
ADR: 6.1,III

Order Now

Specification Test Results

Assay 100%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR