4-Fluorobenzenesulphonyl chloride ≥98%

Supplier: BeanTown Chemical

Synonyms: 4-Fluorobenzene-1-sulphonyl chloride

127015-100G 127015-25G
BT127015-100GEA 155 USD
BT127015-100G BT127015-25G
4-Fluorobenzenesulphonyl chloride ≥98%
4-Fluorobenzenesulphonyl chloride

CAS: 349-88-2; EC No: 206-493-0; MDL No: MFCD00007438 UN No: UN3261; Haz Class: 8; Packing Group: II Crystalline; Molecular Formula: C6H4ClFO2S; MW: 194.61 Melting Point: 29-31°; Boiling Point: 95-96°/2 mmHg; Flash point: 110°C (230°F) Moisture Sensitive


  • Moisture Sensitive
  • Crystalline

Formula: C₆H₄ClFO₂S
Boiling Pt: 95…96 °C/2 mmHg
Melting Pt: 29…31 °C
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00007438
CAS Number: 349-88-2
EINECS: 206-493-0
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR