Nitric acid 68.0 - 70.0% ACS
Supplier: BeanTown Chemical
127005-1LB
127005-7LB
BT127005-1LBEA
53
USD
BT127005-1LB
BT127005-7LB
Nitric acid 68.0 - 70.0% ACS
Nitric acid
CAS: 7697-37-2; EC No: 231-714-2; MDL No: MFCD00011349; RTECS: QU5775000
UN No: UN2031; Haz Class: 8 (5.1); Packing Group: II
Liquid; Molecular Formula: HNO3; MW: 63.01
Boiling Point: 120.5°
Density (g/mL): 1.413
- RTECS: QU5775000
- Liquid
Formula:
HNO₃ Boiling Pt: 120.5 °C Density: 1.413 g/mL |
MDL Number:
MFCD00011349 CAS Number: 7697-37-2 EINECS: 231-714-2 UN: 2031 ADR: 8,II |
Specification Test Results
Assay | 68-70% |
Color (APHA) | ≤10 |
Residue after ignition | ≤5ppm |
Chloride (Cl) | ≤0.5ppm |
Arsenic (As) | ≤0.01ppm |
Sulfate (SO4) | ≤1ppm |
Heavy metals (as Pb) | ≤0.2ppm |
Iron (Fe) | ≤0.2ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...