Zinc phosphate hydrate, Technical Grade

Supplier: BeanTown Chemical

126980-2KG 126980-500G
BT126980-2KGEA 86 USD
BT126980-2KG BT126980-500G
Zinc phosphate hydrate, Technical Grade
Zinc phosphate hydrate

CAS: 7779-90-0; EC No: 231-944-3; MDL No: MFCD00036282; RTECS: TD0590000 UN No: UN3077; Haz Class: 9; Packing Group: III -325 Mesh Powder; Linear Formula: Zn3(PO4)2·xH2O; MW: 386.11 (anhydrous)


  • RTECS: TD0590000
  • -325 Mesh Powder

Formula: O₈P₂Zn₃·xH₂O
MW: 386.08 g/mol (anhy)
MDL Number: MFCD00036282
CAS Number: 7779-90-0
EINECS: 231-944-3
ADR: 9 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR