Cerium(IV) oxide 20% (w/w) in water, colloidal dispersion
Supplier: BeanTown Chemical
Synonyms:
Ceric oxide, Cerium dioxide
126975-100G
126975-2KG
126975-500G
BT126975-100GEA
34
USD
BT126975-100G
BT126975-2KG
BT126975-500G
Cerium(IV) oxide 20% (w/w) in water, colloidal dispersion
Cerium(IV) oxide
CAS: 1306-38-3; EC No: 215-150-4; MDL No: MFCD00010933
UN No: UN3264; Haz Class: 8; Packing Group: III
0.01-0.02 Micron Particles, in Liquid, CeO2 mol/mol 0.2 nitrate, pH 1.5; Molecular Formula: CeO2; MW: 172.12
Boiling Point: 100
Density (g/mL): 1.22
- 0.01-0.02 Micron Particles, in Liquid, CeO2 mol/mol 0.2 nitrate, pH 1.5
Formula:
CeO₂ Boiling Pt: 100 °C Density: 1.22 g/mL |
MDL Number:
MFCD00010933 CAS Number: 1306-38-3 EINECS: 215-150-4 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...