Sodium hydroxide ≥99.99% (trace metals basis)

Supplier: BeanTown Chemical

126950-100G 126950-25G 126950-5G
BT126950-100GEA 113.49 USD
BT126950-100G BT126950-25G BT126950-5G
Sodium hydroxide ≥99.99% (trace metals basis)
Sodium hydroxide

CAS: 1310-73-2; EC No: 215-185-5; MDL No: MFCD00003548; RTECS: WB4900000 UN No: UN1823; Haz Class: 8; Packing Group: II Pellets; Linear Formula: NaOH; MW: 40.00 Melting Point: 318°; Boiling Point: 1390° Density (g/mL): 2.13 Air Sensitive , Hygroscopic


  • RTECS: WB4900000
  • Air Sensitive, Hygroscopic
  • Pellets

Formula: NaOH
Boiling Pt: 1390 °C
Melting Pt: 318 °C
Density: 2.13 g/mL
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1823
ADR: 8,II

Order Now

Specification Test Results

Assay 99.99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR