Acetone-[D₆] ≥100% (Isotopic)
Supplier: BeanTown Chemical
126945-10EACH
126945-2EACH
BT126945-10EAEA
588.28
USD
BT126945-10EA
BT126945-2EACH
Acetone-[D₆] ≥100% (Isotopic)
Acetone-[D6]
CAS: 666-52-4; EC No: 211-563-9; MDL No: MFCD00044635
UN No: UN1090; Haz Class: 3; Packing Group: II
Liquid, in prescored ampules, 0.75ml/ampule ; Linear Formula: CD3COCD3; MW: 64.12
Melting Point: -94°; Boiling Point: 55.5°; Flash point: -19°C (-2°F)
Density (g/mL): 0.872; Refractive Index: 1.355
- Refractive index: 1.355
- Liquid, in prescored ampules, 0.75ml/ampule
Formula:
D₃CC(O)CD₃ Boiling Pt: 55.5 °C Melting Pt: –94 °C Density: 0.872 g/mL Flash Pt: –19 °C (–2 °F) |
MDL Number:
MFCD00044635 CAS Number: 666-52-4 EINECS: 211-563-9 UN: 1090 ADR: 3,II |
Specification Test Results
Assay | 100% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...