Zirconium(IV) ethoxide ≥99%

Supplier: BeanTown Chemical

126860-10G 126860-50G
BT126860-10GEA 130 USD
BT126860-10G BT126860-50G
Zirconium(IV) ethoxide ≥99%
Zirconium(IV) ethoxide

CAS: 18267-08-8; MDL No: MFCD00015654 UN No: UN3181; Haz Class: 4.1; Packing Group: III Crystalline; Linear Formula: Zr(OCH2CH3)4; MW: 271.47 Melting Point: 171-173°; Boiling Point: 235°/5 mmHg Moisture Sensitive


  • Moisture Sensitive
  • Crystalline

Formula: C₈H₂₀O₄Zr
MW: 271.47 g/mol
Boiling Pt: 235 °C/5 mmHg
Melting Pt: 171…173 °C
MDL Number: MFCD00015654
CAS Number: 18267-08-8
UN: 3181
ADR: 4.1,III

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR